Alfa Aesar™ trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl, 97%

Manufacturer: Alfa Aesar™ H5516806

Quantity 5g

 View more versions of this product

Catalog No. AAH5516806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 118164-51-5
    Molecular Formula C23H34F2
    Formula Weight 348.52
    MDL Number MFCD13182310
    Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl

    Specifications

    Formula Weight 348.52
    Molecular Formula C23H34F2
    MDL Number MFCD13182310
    CAS 118164-51-5
    Melting Point 45° to 47°C
    Quantity 5g
    Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl
    Assay 97%

    Description

    Applications
    It is used as flavor and fragrance intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

    Solubility
    Slightly soluble in water.

    Notes
    Reacts with strong oxidizing agents. Keep at room temperature.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit