Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ Transferpette™ -8/-12 electronic Multichannel Pipette - TRADE-UP PROMO
Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs. PROMO - Save when you upgrade your old pipette to the Brand™ Transferpette™ pipette.
Supplier: BrandTech™ 705424TU
Description
- Choice of 8 or 12 channel models
- Clear, legible LCD screen
- Intuitive interface with user-friendly desing allows easy operation
- Soft touch pipetting key and adjustable finger rest provides added comfort
- Rechargeable battery provides over 4000 pipetting cycles between charges
- Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
- Easily maintained
Operating modes
- Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
- Mixing of samples (PIPmix Mode): program for mixing liquids
- Reverse Pipetting (revPIP Mode): program designed for greater accuracy
- Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
- Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
Specifications
±1.6μL | |
2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip, 5 to 200μL tip | |
8 | |
Step Based | |
10 to 200 μL | |
Ergonomic | |
0.2μL | |
1 Year |
Rechargeable | |
Transferpette™ -8/-12 Electronic | |
Transferpette™ | |
Electronic Pipette | |
LCD | |
Battery, power supply unit, 120VAC adapter with charging cable, one pipette tip sample, operating manual, pipette stand, tip-box SL filled with appropriate tip, two tip refill inserts, reagent reservoir and five operating modes | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction