Learn More
BrandTech™ Transferpette™ S Single Channel Adjustable Pipet - TRADE-UP PROMO
Accurate and precisise with maximum versatility and reliable performance with Easy Calibration™ tech. Simple one-handed operation with adjustable brake to protect from accidental volume changes. PROMO - Save when you upgrade your old pipette to the Brand™ Transferpette™ pipette.
Manufacturer:  BrandTech™ 704780TU
 View more versions of this product
Catalog No. 14-389-105TU
Add to cart
Description
Description
Single channel model is available in fixed volume and digital models in volume ranges from 0.1µL to 10mL.
  • Centralized pipetting button with optimized stroke length
  • Adjustable brake to fix volume range
  • Visible four-digit volume display
  • Compatible with universal tips (0.1-1µL requires nano-cap tips)
  • Repeatedly autoclavable at 121°C/250°F(20min.)
Specifications
Specifications
±0.6%
CE, IVD
Digital
Demanding laboratory applications
Performance certificate, operating manual, shelf mount, and three year warranty.
Single channel pipet
D-1000
Autoclavable
50 to 1000μL tip, 50 to 1250μL tip, 50 to 1000μL tip
Curved
50 to 1000μL tip, 50 to 1250μL tip, 50 to 1000μL tip
1μL
3 Years
100 to 1000 μL