Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin, Mouse anti-Human, Alexa Fluor 488, Clone: TF/3001, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279912AF488

Catalog No. NB012408


Add to cart

Description

Description

Transferrin Monoclonal antibody specifically detects Transferrin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Transferrin
TF/3001
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TF
Recombinant fragment (around aa 311-445) of human Transferrin protein (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Mouse
IgG1 Kappa
0.1 mL
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit