Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transferrin, Mouse anti-Human, Clone: TF/3001, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279778100UG
Description
Transferrin Monoclonal specifically detects Transferrin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Transferrin | |
Monoclonal | |
0.2 mg/mL | |
ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P02787 | |
TF | |
Recombinant fragment (around aa 311-445) of human Transferrin protein (exact sequence is proprietary) | |
100 μg | |
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers | |
7018 | |
Store at 4°C. | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
TF/3001 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction