Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transferrin, Mouse anti-Human, Clone: TF/3001, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP27977820UG
Description
Transferrin Monoclonal antibody specifically detects Transferrin in Human Samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array.Specifications
Transferrin | |
TF/3001 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Purified | |
TF | |
Recombinant fragment (around aa 311-445) of human Transferrin protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C. | |
Monoclonal | |
Human |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
0.2 mg/mL | |
ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P02787 | |
Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
Mouse | |
IgG1 Kappa | |
20 ug | |
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers | |
Primary | |
7018 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title