Learn More
Transferrin Rabbit anti-Bovine, Polyclonal, MP Biomedicals™
Rabbit Polyclonal Antibody
Manufacturer:  MP Biomedicals, Inc 0855273
Catalog No. MP0855273
Add to cart
Description
Description
  • Product is the lyophilized powder of rabbit antiserum to bovine transferrin and buffer salts.
  • The antiserum is prepared by hyperimmunizing rabbits with highly purified bovine transferrin; antiserum is delipidated and made specific by solid-phase absorption
  • This product is suitable for use as a precipitating antibody. It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques.
Specifications
Specifications
Transferrin
Unlabeled
Product is the lyophilized powder of rabbit antiserum to bovine transferrin and buffer salts.
Lyophilized powder
2mL
Store at 2-8°C.
Polyclonal
Bovine
Immunohistochemistry, ELISA
Unconjugated
0.02M sodium phosphate, 0.14M sodium chloride,pH 7.3
Rabbit
RUO
Primary
Reconstitute product with 2mL of deionized or distilled water.gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material.