Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1780970.5MG

Catalog No. NBP17809705

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TransferrinPolyclonalantibody specifically detects Transferrin in Human samples.
Specifications

Specifications

Transferrin
Unconjugated
0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
TF
Human Transferrin
Immunogen affinity purified
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Primary
7018
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:500-1:2500, Immunohistochemistry-Paraffin 1:10-1:500
P02787
Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Rabbit
IgG
0.5mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Analysis by SDS-PAGE was used to determine that this preparation is substantially free of aggregates and shows a banding pattern consistent with purified Rabbit IgG.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit