Learn More
Globe Scientific TransferTop™ Urine Collection and Transfer Container
Globe Scientific TransferTop™ Urine Collection and Transfer Container feature an integrated transfer mechanism for closed system transfer of urine samples directly from the cup to a vacuum sample tube.
Manufacturer:  Globe Scientific 3856L
Catalog No. 22-171-724
Add to cart
Description
Description
  • Requires no open transferring of samples for reduced spills, improved lab hygiene, and limited exposure to dangerous samples
  • Reduces risk of contamination for improved test results
  • Directly transfers samples to a tube suitable for analysis and transport
  • Works with most vacuum urine tubes
  • Provides consistent sample transfer volumes when used with like vacuum tubes
  • Improves efficiencies and sample throughput when compared to traditional open systems
Order Info
Case of 300
Specifications
Specifications
Specimen Container
120mL
Yellow
Natural
Sterile
4oz.
Polypropylene
Screwcap
300