Learn More
Thermo Scientific™ TransformAid Bacterial Transformation Kit
Kit for quick and convenient preparation of competent E.coli cells for guaranted transformation efficiencies of more than 107 transformants per µg of plasmid DNA.
$50.30
Specifications
For Use With (Application) Routine cloning experiments.Blunt-end cloning.TA cloning.
View More Specs
Products 1
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
FERK2710
View Documents
Thermo Scientific™
K2710
Each for $50.30
Add to cart
Description
Description
Employ a novel method for rapid preparation of chemically-competent E. coli cells from overnight bacterial cultures or bacterial colonies with the Thermo Scientific TransformAid™ Bacterial Transformation Kit. The key component of this system is the unique T-solution, which produces competent cells in a few easy steps. The quick and convenient procedure guarantees transformation efficiencies of more than 107 transformants per µg of plasmid DNA, ideal for routine cloning experiments. The option to use bacterial colonies for preparation of competent cells adds flexibility to any cloning experiment.
The TransformAid Bacterial Transformation Kit can be used with most E. coli strains commonly used for cloning: JM107, JM109, XL1-Blue, SURE, TOPP2, W3110, NM527, AD494, CJ236, GM2163, C600, BL21, M15, NM522, HB101, ER2267.
Highlights
  • Efficient – more than 107 transformants per µg of plasmid DNA
  • Practical – E. coli competent cells can be prepared from an overnight culture or from bacterial colonies
  • Fast – less than 1 hour from the overnight bacterial culture to cell plating. Starting from bacterial colonies, the procedure requires only 2.5 hours
  • Easy – all procedures are performed on ice on your bench. Brief centrifugations are carried out in regular microcentrifuges
Applications
  • Routine cloning experiments
  • Blunt-end cloning
  • TA cloning
Includes
  • C-Medium
  • T-Solution (A)
  • T-Solution (B)
  • Detailed Protocol
Note
Competent cells prepared with TransformAid Bacterial Transformation Kit are suitable for direct-use only. Freezing down and storage at -70°C is not recommended.
Specifications
Specifications
Routine cloning experiments.Blunt-end cloning.TA cloning.