Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Transforming Growth Factor-b (TGF-b), Human Natural

Corning™ Transforming Growth Factor-beta (TGF-beta), human natural is a multifunctional protein that plays a central role in the regulation of cell growth and differentiation with either stimulatory or inhibitory effects.

Manufacturer:  Corning™ 356040

 View more versions of this product

Catalog No. MT356040


Add to cart

Description

Description

Formulation

Frozen in 36% acetonitrile-0.1% trifluoroacetic acid
Caution: TGF-ß absorbs to glass and plastic surfaces (instructions for dilution are provided with the product)

Source

Human platelets. NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody

Molecular Weight

25 kD

Quality

  • Purity: >95% by SDS-PAGE
  • Tested for ability to inhibit DNA synthesis of Mv 1Lu mink lung cells (ATCC CCL-64)
  • Tested and found negative for bacteria, fungi and mycoplasma
    • Tip

      Recommended concentration: 10-50 pg/mL of medium depending on cell type and application.

      Preparation and Storage

      Stable for at least three months at -70°C (if diluted according to instructions)

      Storage

      Shelf life: ≥ 3 months from date of shipment at -70°C

Specifications

Specifications

25 kD
2 µg/Container, 5/Pk.
≥ 3 months from date of shipment (at -70°C)
10 µg
>95% by SDS-PAGE
Human Platelets
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit