Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS670B1

 View more versions of this product

Catalog No. MS670B1


Add to cart

Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody.

TGFα (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Host Species: Mouse

Clone: MF9

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Rat and Mouse. Others not known.

Epitope: C-terminal

Immunogen: A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFα

Molecular Weight: 6kDa

Positive Control: Recombinant TGFα protein, A2058 cells

Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with EGF and the neuropeptide synenkephalin. Ab-1 is excellent for staining of formalin-fixed, paraffin-embedded tissues. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Transforming Growth Factor alpha (TGFalpha) Ab-1
Monoclonal
200μg/mL
Concentrated
A 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha
500μL
Signal Transduction
Primary
Bovine, Human, Murine, Rat
Western Blot
MF9
Biotin
Murine
IgG2a kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit