Learn More
Transforming Growth Factor α (TGFα) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Transforming Growth Factor α (TGFα) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS670P1
Description
TGFα (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.Host Species: Mouse
Clone: MF9
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Rat and Mouse. Others not known.
Epitope: C-terminal
Immunogen: A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFα
Molecular Weight: 6kDa
Positive Control: Recombinant TGFα protein, A2058 cells
Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with EGF and the neuropeptide synenkephalin. Ab-1 is excellent for staining of formalin-fixed, paraffin-embedded tissues. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody.
Recommended for:
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Transforming Growth Factor alpha (TGFalpha) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Signal Transduction | |
Bovine, Human, Mouse, Rat | |
IgG2a κ |
Western Blot | |
MF9 | |
Unconjugated | |
A 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.