Learn More
ADS Biotec™ Optimase™ DNA Polymerase
Ideal for the detection of low levels of mutation in wild type/mutant DNA heterogenous samples and other high-fidelity PCR applications
Manufacturer:  ADS Biotec™ 703005
 View more versions of this product
Catalog No. 703005
  Check Availability
Add to cart
Specifications
Description
Optimase™ Polymerase is ideal for low level detection of mutations in wild type/mutant DNA heterogenous samples. It is suited for use in amplicon-specific PCR protocols and exhibits high thermal stability.
Optimase™ Polymerase checks each nucleotide during DNA synthesis to excise mismatched nucleotides in the 3´ to 5´ direction. OptimasePolymerase proofreading enzyme also enables the removal of unpaired 3´ overhanging nucleotides to create blunt ends.
  • Thermostable proofreading DNA polymerase featuring efficient 3 ft.5 ft. exonuclease activity
  • Offers superior performance in single nucleotide polymorphism (SNP) and mutation discovery studies
  • Can be used in amplicon-specific PCR protocols developed by Optimase ProtocolWriter™ (please visit www.mutationdiscovery.com for details)
  • Manufactured under ISO 9001 Quality Management System 
  • Quality-control tested for DNA polymerase activity, endonuclease contamination, and fidelity tests
  •  Provided in storage buffer
Description & Specifications
Specifications
PCR applications the require high-fidelity and exhibits supreior performance in single nucleotide polymorphism (SNP) and mutation studies.   
  • Optimase™ Polymerase: 1 x 100 Units
  • 10X Reaction Buffer (Mg free): 1 x 1.5mL
  • 25mM MgSO4: 1 x 1.0mL
100U
-20°C
Optimase DNA Polymerase