Transgenomic™ Surveyor™ Mutation Detection Kit for Standard Gel Electrophoresis
Detects all types of point mutations and small insertions or deletions
Manufacturer: Transgenomic, Inc. 706020
 View all options for this product
Catalog No. 706020
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    For Use With (Application) SNP and mutation detection
    For Use With (Equipment) Standard Gel electrophoresis
    Includes Surveyor Nuclease S, surveyor Nuclease enhancer S, 10 x survey or Nuclease Reaction Buffer, stop solution, control plasmids, control primers
    Quantity 100 Reactions
    Provides a simple and robust method to scan DNA fragments for known and unknown mutations and single nucleotide polymorphisms. Mutations can be detected in four steps: PCR amplification of mutant and reference DNA, hybridization to form hetero- and homoduplexes, digestion with Surveyor Nuclease, and separation by standard agarose or polyacrylamide electrophoresis.
    ·    Utilizes Surveyor Nuclease, which specifically cleaves DNA heteroduplexes at mismatch sites
    ·    Suitable for use with large fragments up to several kb
    ·    Can detect multiple mutations in a single fragment
    ·    Ideal for the verification of the presence or absence of mutations or the localization of mutations to minimize the extent of DNA sequencing
  • Description & Specifications
Related Products
Product Content Feedback