Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Transglutaminase 2/TGM2 Colorimetric Assay Kit

Applications: Functional

Manufacturer:  Novus Biologicals™ NBP1370082PLATES

Catalog No. NBP1370082P


Add to cart

Description

Description

  • Marker: Tissure Specific Transglutaminase Colorimetric Microassay
  • Gene: TGM2
Specifications

Specifications

2 vials (Lyophilized powder)
2 plates
Transglutaminase 2/TGM2 Colorimetric Assay Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only.