Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transglutaminase 2/TGM2 Mouse anti-Human, Canine, Guinea Pig, Rabbit, Clone: CUB7402, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Transglutaminase 2/TGM2 |
---|---|
Clone | CUB7402 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Transglutaminase 2/TGM2 Monoclonal specifically detects Transglutaminase 2/TGM2 in Human, Canine, Guinea Pig, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Transglutaminase 2/TGM2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2 | |
TGM2 | |
IgG1 | |
Affinity Purified |
CUB7402 | |
Monoclonal | |
Mouse | |
Apoptosis, Immune System Diseases, Immunology, Signal Transduction | |
7052 | |
BALB/C mice were injected with purified guinea pig liver TGase II. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title