Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Transglutaminase II Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-1, Mouse Monoclonal Antibody.

$219.00 - $1012.00

Specifications

Antigen Transglutaminase II Ab-1
Clone CUB 7402
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS224B
View Documents
Thermo Scientific™
MS224B
Biotinylated Ab with BSA and Azide 1mL
Each for $995.00
Add to cart
 
MS224B0
View Documents
Thermo Scientific™
MS224B0
Biotinylated Ab with BSA and Azide 100μL
Each for $232.00
Add to cart
 
MS224B1
View Documents
Thermo Scientific™
MS224B1
Biotinylated Ab with BSA and Azide 500μL
Each for $628.00
Add to cart
 
MS224P
View Documents
Thermo Scientific™
MS224P
Purified Ab with BSA and Azide 1mL
Each for $1,012.00
Add to cart
 
MS224P0
View Documents
Thermo Scientific™
MS224P0
Purified Ab with BSA and Azide 100μL
Each for $219.00
Add to cart
 
MS224P1
View Documents
Thermo Scientific™
MS224P1
Purified Ab with BSA and Azide 500μL
Each for $627.50
Add to cart
 
MS224P1ABX
View Documents
Thermo Scientific™
MS224P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $651.00
Add to cart
 
MS224PABX
View Documents
Thermo Scientific™
MS224PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $869.00
Add to cart
 
MS224R7
View Documents
Thermo Scientific™
MS224R7
Ready-to-use for IHC (UVLP) 7mL
Each for $453.00
Add to cart
 
MS224R1
View Documents
Thermo Scientific™
MS224R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-1, Mouse Monoclonal Antibody.

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: CUB 7402

Isotype: IgG1/κ

Species Reactivity: Human, Dog, Rabbit and Guinea Pig. Does not react with mouse and rat. Others not known.

Epitope: aa 447-478

Immunogen: Purified guinea pig liver TGase II

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-1 inhibits the TGase II activity.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Affinity Purification
  • Biological Blockade

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Transglutaminase II Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Neuroscience
Purified guinea pig liver TGase II
CUB 7402
Monoclonal
RUO
Primary
IgG2a kappa
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit