Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Transglutaminase II Ab-4, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-4, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB060P0

 View more versions of this product

Catalog No. RB060P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Transglutaminase II Ab-4, Rabbit Polyclonal Antibody.

TGase II is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant full-length human TGase II

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-4 is excellent for staining of endothelial cells in murine tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Transglutaminase II Ab-4
Polyclonal
Unconjugated
Concentrated
Recombinant full length human TGase II
100μL
Neuroscience
Primary
Human, Murine
Immunoprecipitation, Western Blot
1mg/mL
Purified Ab with BSA and Azide
Rabbit
IgG
RUO
Primary
Polyclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit