Learn More
AVANTOR MASTERFLEX (VWR LLC) Flowline LU84-5101 Ultrasonic Level Transmitter; 32.8' Range, 2
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4331265
Encompass_Preferred
EchoSpan™, a general-purpose two-wire ultrasonic transmitter, provides noncontact level measurement up to 32 ft or 10 m, and is ideally suited for challenging ultrapure, corrosive or waste liquids. Push-button calibrated, the transmitter is broadly selected for atmospheric bulk storage, day tank and waste sump applications. Media examples include wastewater and sodium hydroxide.
Catalog No. 50-630-877