Learn More
AVANTOR MASTERFLEX (VWR LLC) Flowline LU83-5101 Large Tank Transmitter, 4 to 20 mA, 8
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX4326000
EchoSpan™, a general-purpose two-wire ultrasonic transmitter, provides noncontact level measurement up to 32 ft or 10 m, and is ideally suited for challenging ultrapure, corrosive or waste liquids. Push-button calibrated, the transmitter is broadly selected for atmospheric bulk storage, day tank and waste sump applications. Media examples include wastewater and sodium hydroxide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.