Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Transport Vials, 10mL

A comprehensive range of premium quality, durable polypropylene vials that coordinate with capping options for either storage or transport.

Manufacturer:  Starplex™ Scientific V102 NL

 View more versions of this product

Catalog No. 14-375-437


Add to cart

Description

Description

Leak resistant, graduated 10ml vials are available in clarified polypropylene for easy visual inspection or amber for light sensitive testing. Flat, white, o-ring style screw caps provided separately in the case are ideal for transport. 

The 10mL white capped vial is also available in a sterile peel pouch.

Specifications

Specifications

Polypropylene
10 mL
Cap
Specimen collection/transportation
Non-sterile
White
Translucent
No Label
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit