Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transthyretin/Prealbumin Mouse anti-Human, Clone: TTR/4292, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30774220UG

 View more versions of this product

Catalog No. NB122958


Add to cart

Description

Description

Transthyretin/Prealbumin Monoclonal specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Transthyretin/Prealbumin
TTR/4292
Unconjugated
10 mM PBS with 0.05% BSA
ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin
Recombinant human full-length protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG2c Kappa
20 μg
Cancer
Primary
7276
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit