Learn More
TRAP1 Mouse anti-Human, Mouse, Rat, HRP, Clone: 3H4-2H6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259343H
Catalog No. NBP259343H
Add to cart
Description
Description
TRAP1 Monoclonal antibody specifically detects TRAP1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
TRAP1
3H4-2H6
Western Blot
Purified
TRAP1
Purified recombinant TRAP1
Protein G purified
Membrane Trafficking and Chaperones, Stem Cell Markers
Primary
10131
Human, Mouse, Rat
Western Blot
HRP
PBS with No Preservative
HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein
Mouse
IgG1 Kappa
100 ul
Store at 4C in the dark.
Monoclonal
Detects 75kDa.