Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRAPPC10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577021
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
TRAPPC10, also known as TMEM1 (transmembrane protein 1), EHOC1 (epilepsy holoprosencephaly candidate 1 protein) or GT334, is a widely expressed 1,259 amino acid protein that may function in vesicular transport. Despite its name, TRAPPC10 does not contain transmembrane domains. It is the human ortholog of the yeast Trs130 protein and its structure and function appears to be conserved. Localizing to the cis-Golgi apparatus, TRAPPC10 is believed to be involved in transport from the endoplasmic reticulum (ER) to the Golgi functioning as a component of the multisubunit transport protein particle (TRAPP) complex. Mutations in the gene encoding TRAPPC10 may be involved in autoimmune polyglandular disease type 1 or Unverricht-Lundborg disease, an autosomal recessive type of progressive myoclonic epilepsy.
Specifications
TRAPPC10 | |
Polyclonal | |
Unconjugated | |
TRAPPC10 | |
B230307C21Rik; EHOC1; EHOC-1; epilepsy holoprosencephaly candidate 1 protein; epilepsy holoprosencephaly candidate-1 protein; Gm870; GT334; Protein GT334; TMEM1; trafficking protein particle complex 10; trafficking protein particle complex subunit 10; trafficking protein particle complex subunit 10; LOW QUALITY PROTEIN: trafficking protein particle complex subunit 10; trafficking protein particle complex subunit 130; trafficking protein particle complex subunit TMEM1; transmembrane protein 1; transport protein particle subunit TMEM1; TRAPP 130 kDa subunit; TRAPP subunit TMEM1; trappc10; TRS130; TRS30; wu:fc21a02 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216131, 309678, 7109 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P48553, Q3TLI0 | |
TRAPPC10 | |
Recombinant full length Human TRAPPC10. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction