Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548747

Catalog No. PA548747

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Prevents transcriptional repression and induction of cell death by ENO1. May play a role in vesicular transport from endoplasmic reticulum to Golgi.
Specifications

Specifications

TRAPPC2
Polyclonal
PBS with 0.09% sodium azide
P0DI81
Trafficking protein particle complex subunit 2, Sedlin, TRAPPC2, SEDL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6399
Western Blot
Unconjugated
TRAPPC2
Liquid
TRAPPC2
KLH conjugated synthetic peptide between 11-39 amino acids from the N-terminal region of human TRAPPC2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit