Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAPPC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$166.00 - $405.00
Specifications
Antigen | TRAPPC2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121527
![]() |
Thermo Scientific
PA5121527 |
100 μL |
Each for $405.00
|
|
PIPA5121528
![]() |
Thermo Scientific
PA5121528 |
20 μL |
Each for $166.00
|
|
Description
TRAPPC2 Polyclonal antibody specifically detects TRAPPC2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
TRAPPC2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
TRAPPC2 | |
P0DI81 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
TRAPPC2 | |
Primary | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title