Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550144

Catalog No. PA550144


Add to cart

Description

Description

The antibody detects endogenous levels of total TRAPPC3 protein.

TRAPPC3 is a component of the TRAPP complex, which isinvolved in tethering of transport vesicles to the cis-Golgimembrane.
Specifications

Specifications

TRAPPC3
Polyclonal
Unconjugated
TRAPPC3
Liquid
TRAPPC3
Synthesized peptide derived from internal of human TRAPPC3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O43617, O55013, Q5U1Z2
BET3
Rabbit
IgG
100 μL
-20°C
Primary
27095, 27096, 362599
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit