Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRAPPC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575940
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
TRAPPC5 (trafficking protein particle complex 5), also known as TRS31, is a 181 amino acid protein belonging to the TRAPP (transport protein particle) small subunits family and the BET3 subfamily. Encoded by a gene that maps to human chromosome 19p13.2, TRAPPC5 is part of the multisubunit TRAPP tethering complex, which acts as a GTP exchange factor. Evolutionarily conserved, TRAPPC5 plays a role in protein binding,vesicle-mediated transport and nucleotide exchange stimulation. TRAPPC5 also performs guanine nucleotide exchanger factor (GEF) functions both in vitro and in vivo. Localizing to the Golgi apparatus, TRAPPC5 is essential for endoplasmic reticulum (ER)-to-Golgi and intra-Golgi vesicle trafficking in yeast, as well as additional transport events in mammals, such as post-Golgi trafficking.
Specifications
TRAPPC5 | |
Polyclonal | |
Unconjugated | |
TRAPPC5 | |
4021401A16Rik; RGD1563971; trafficking protein particle complex 5; trafficking protein particle complex subunit 5; Trappc5; TRS31 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
126003, 363858, 66682 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IUR0, Q9CQA1 | |
TRAPPC5 | |
Synthetic peptide corresponding to amino acids 68-113 of Human TRAPPC5. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction