Learn More
Trastuzumab Rabbit anti-Multi-Species, Alexa Fluor 594, Clone: 2261E, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB95471T100UG
Catalog No. FAB9547T100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Trastuzumab
2261E
Flow Cytometry 0.25-1 ug/10^6 cells
Supernatant
Full-length Trastuzumab
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Multi-Species
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide
Rabbit
IgG
100 ug
Primary
Detects Trastuzumab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed.