Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trazodone Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
25332-39-2
C19H23Cl2N5O
408.33
OHHDIOKRWWOXMT-UHFFFAOYSA-N
hydrogen 2-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one chloride
[H+].[Cl-].ClC1=CC=CC(=C1)N1CCN(CCCN2N=C3C=CC=CN3C2=O)CC1
25332-39-2
C19H23Cl2N5O
408.33
OHHDIOKRWWOXMT-UHFFFAOYSA-N
hydrogen 2-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one chloride
[H+].[Cl-].ClC1=CC=CC(=C1)N1CCN(CCCN2N=C3C=CC=CN3C2=O)CC1

T1149, 25332-39-2, C19H22ClN5O•HCl

Spectrum™ Trazodone Hydrochloride, USP is used to treat depression. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.002
Loss on Drying 0.005
Quantity 5 g
CAS Max % 1
Percent Purity 98 to 102%
Grade USP