Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRBC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549387
Description
TRBC1 Polyclonal antibody specifically detects TRBC1 in Human samples. It is validated for Western Blot.Specifications
TRBC1 | |
Polyclonal | |
Unconjugated | |
TRBC1 | |
BV05S1J2.2; T cell receptor beta constant 1; TCRB; TCRBC1; TRBC1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
28639 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P01850 | |
TRBC1 | |
KLH conjugated synthetic peptide between 104-136 amino acids from the Central region of human TRBC1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction