Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRBP Mouse anti-Human, Clone: PCRP-TARBP2-1E5, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | TRBP |
---|---|
Clone | PCRP-TARBP2-1E5 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122144
|
Novus Biologicals
NBP30733520UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
TRBP Monoclonal specifically detects TRBP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
TRBP | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
LOQS, RISC-loading complex subunit TARBP2, TAR (HIV) RNA binding protein 2, TAR (HIV) RNA-binding protein 2, TAR (HIV) RNA-binding protein TRBP1, TAR (HIV-1) RNA binding protein 2, TAR RNA binding protein 2, TAR RNA-binding protein 2, trans-activation responsive RNA-binding protein, Trans-activation-responsive RNA-binding protein, TRBP, TRBP1, TRBP2 | |
Recombinant full-length human TRBP protein | |
Primary | |
Store at 4°C. |
PCRP-TARBP2-1E5 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
6895 | |
IgG2a | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title