Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRBP, Rabbit anti-Human,Mouse,Rat,Bovine,Primate,Zebrafish, DyLight 550, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056471R

Catalog No. NB003609

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TRBP Polyclonal antibody specifically detects TRBP in Human, Mouse, Rat, Bovine, Primate, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

TRBP
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
LOQS, RISC-loading complex subunit TARBP2, TAR (HIV) RNA binding protein 2, TAR (HIV) RNA-binding protein 2, TAR (HIV) RNA-binding protein TRBP1, TAR (HIV-1) RNA binding protein 2, TAR RNA binding protein 2, TAR RNA-binding protein 2, trans-activation responsive RNA-binding protein, Trans-activation-responsive RNA-binding protein, TRBP, TRBP1, TRBP2
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat, Bovine, Primate, Zebrafish
Western Blot
Western Blot
Affinity Purified, Conjugated
TARBP2
A portion of amino acids 150-200 of human TRBP2 was used as the immunogen.
Immunogen affinity purified
RUO
Primary
Human isoform a is a 366 amino acid protein, and human isoform b is a 345 amino acid protein.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit