Learn More
Tocris Bioscience TRC 051384
Inducer of heat shock protein Hsp70
Manufacturer: Tocris Bioscience 4734/10
Description
Inducer of heat shock protein Hsp70; reduces stroke-associated neuronal injury and increases survival in a rat model of transient ischemic stroke. Activates heat shock factor-1 and results in elevated chaperone and anti-inflammatory activity. Enhances Hsp72 expression in neurons and glial cells.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
TRC 051384 | |
>98% | |
10mg |
TRC 051384 | |
465.54 |