Learn More
XenoTech,LLC TREATED MIC SALINE CYNO 0.5 ML
SDP
Supplier:  XenoTech,LLC P2073
Encompass_Preferred
Pooled saline-treated Cynomolgus monkey liver microsomes, male, 0.5 mL @ 10 mg/mL (5 mg)
Catalog No. 50-142-985