Learn More
Biotium Trefoil factor 2 / TFF2(GE16C), 1mg/mL
Supplier:  Biotium BNUM306050
Encompass_Preferred
Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. These peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain.This antibody is available purified, with BSA and azide (0.2 mg/mL) or purified, BSA- and azide-free (1 mg/mL).
Catalog No. 50-006-16492