Learn More
Ansell™ Trellchem™ Bayonet Cuff Ring System
Complete one ring system for all glove combinations
Manufacturer:  Ansell™ 487010605
Catalog No. 19-810-212
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Quick and simple glove exchange
  • Safe and easy to assemble
  • Positions are easy to check
  • Distinct markings allow system to be manuveured in low-light conditions
  • Locking pin for added safety
  • Allow attachment of 4H inner gloves; gloves stay inside outer glove when hand is removed
  • Glove ring inner dia.: 98mm
  • Two Viton™ O-rings and glove compression for optimal sealing effect
Certifications
Meets NFPA 1991 and EN 943-1 and -2
Specifications
Specifications
NFPA 1991, EN 943-1, -2
All glove combinations
Includes: 2 Viton™ O-rings and glove compression for optimal sealing, locking pin for safety