Learn More
Ansell™ Trellchem™ Complete Repair Kits
For repair of minor damage such as small holes or punctures to Trellchem protective suits
Manufacturer:  Ansell™ 487080400
 View more versions of this product
Catalog No. 19-810-215
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
All suits shoulld be inspected after every use or at least once every year including gastightness test, checking the suit material and components for damage and lubricating the zipper.
Includes: Each kit includes eight patches (two sizes), one 125mL can of adhesive (for outside repair), one 250mL bottle of solvent for cleaning, one 8mL bottle of hardener (sufficient for 125mL of adhesive), two brushes, transparent tape on release paper for inside repair, and kit box
Description
Description
Separate kits for each type of Trellchem suit. Follow repair instructions detailed in the manual provided with each suit.
Specifications
Specifications
For repair of HPS level A protection suits
VPS Level A Suits
Eight patches (two sizes), one 125mL can of adhesive, one 250mL bottle of solvent for cleaning, one 8mL bottle of hardener (sufficient for 125mL of adhesive), two brushes, transparent tape on release Paper for inside repair and kit box