Learn More
Ansell™ Trellchem™ Complete Repair Kits
Click to view available options
Product Type:
HPS Level A Suits
Light Protective Suits
Description
Includes
Each kit includes eight patches (two sizes), one 125mL can of adhesive (for outside repair), one 250mL bottle of solvent for cleaning, one 8mL bottle of hardener (sufficient for 125mL of adhesive), two brushes, transparent tape on release paper for inside repair, and kit box
Separate kits for each type of Trellchem suit. Follow repair instructions detailed in the manual provided with each suit.
Specifications
For Use With (Application) | For repair of HPS level A protection suits |
Description | Eight patches (two sizes), one 125mL can of adhesive, one 250mL bottle of solvent for cleaning, one 8mL bottle of hardener (sufficient for 125mL of adhesive), two brushes, transparent tape on release Paper for inside repair and kit box |
Product Type | HPS Level A Suits |