Learn More
Trellchem™ Complete Repair Kits
For repair of minor damage such as small holes or punctures to Trellchem protective suits
Manufacturer:  Trelleborg 487080073
 View more versions of this product
Catalog No. 19-810-218
  Check Availability
Add to cart
Alert:
All suits shoulld be inspected after every use or at least once every year including gastightness test, checking the suit material and components for damage and lubricating the zipper.
Specifications Description & Specifications
Specifications
Super-T Level B Suits
Eight patches (two sizes), one 125mL can of adhesive, one 250mL bottle of solvent for cleaning, one 8mL bottle of hardener (sufficient for 125mL of adhesive), two brushes, transparent tape on release Paper for inside repair and kit box
For repair of HPS level A protection suits