Learn More
Ansell™ Trellchem™ evo Hazardous Protection Suits
Includes Nomex layer in material for additional chemical flash fire protection
Manufacturer:  Ansell™ 477520062
 View more versions of this product
Catalog No. 19-810-024
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Single skin; no overcover required
  • Antistatic resistance built into material; highly abrasion resistant
  • Large, impact-resistant visor and antifog shield
  • Available with attached socks in the suit material or attached nitrile black boots†
  • Includes ventilation system
  • Fully repairable
  • Red
Certifications
Suits with attached socks are NFPA 1991-2005 certified with chemical flash fire and liquefied gas options.
Warranty and Services
Two year
Specifications
Specifications
NFPA 1991-2005
Medium
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system
Red
Attached socks
Nomex™