Learn More
Ansell™ Trellchem™ evo Hazardous Protection Suits
Includes Nomex layer in material for additional chemical flash fire protection
Specifications
Certifications/Compliance NFPA 1991-2005
Color Red
Material Nomex™
View More Specs
Products 12
Catalog Number Mfr. No. Description Size Price Quantity & Availability  
Catalog Number Mfr. No. Description Size Price Quantity & Availability  
19-810-023 View Documents Ansell™
477520061
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-024 View Documents Ansell™
477520062
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-025 View Documents Ansell™
477520063
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-026 View Documents Ansell™
477520064
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-027 View Documents Ansell™
477520065
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-028 View Documents Ansell™
477520066
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-030 View Documents Ansell™
477520062BOOT
Attached Nitrile black boots, large, impact-resistant visor and anti-fog shield, ventilation system Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-029 View Documents Ansell™
477520061BOOT
Attached Nitrile black boots, large, impact-resistant visor and anti-fog shield, ventilation system Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-031 View Documents Ansell™
477520063BOOT
Attached Nitrile black boots, large, impact-resistant visor and anti-fog shield, ventilation system Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-032 View Documents Ansell™
477520064BOOT
Attached Nitrile black boots, large, impact-resistant visor and anti-fog shield, ventilation system X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-033 View Documents Ansell™
477520065BOOT
Attached Nitrile black boots, large, impact-resistant visor and anti-fog shield, ventilation system 2X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-034 View Documents Ansell™
477520066BOOT
Attached Nitrile black boots, large, impact-resistant visor and anti-fog shield, ventilation system 3X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Single skin; no overcover required
  • Antistatic resistance built into material; highly abrasion resistant
  • Large, impact-resistant visor and antifog shield
  • Available with attached socks in the suit material or attached nitrile black boots†
  • Includes ventilation system
  • Fully repairable
  • Red
Certifications
Suits with attached socks are NFPA 1991-2005 certified with chemical flash fire and liquefied gas options.
Warranty and Services
Two year
Specifications
Specifications
NFPA 1991-2005
Red
Nomex™