Trellchem™ evo Hazardous Protection Suits Available on GSA/VA Contract for Federal Government customers only.
Includes Nomex layer in material for additional chemical flash fire protection
Manufacturer: Trelleborg 477520064
 View more versions of this product
Catalog No. 19-810-026
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1991-2005
    Material Nomex™
    Includes Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system
    Color Red
    Size X-Large
  • Description & Specifications
Related Products
Product Content Feedback