Learn More
Trellchem™ evo Hazardous Protection Suits Available on GSA/VA Contract for Federal Government customers only.
Includes Nomex layer in material for additional chemical flash fire protection
Manufacturer:  Trelleborg 477520065
 View more versions of this product
Catalog No. 19-810-027
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1991-2005
Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system
2X-Large
Nomex™
Red