Trellchem™ evo Hazardous Protection Suits
Includes Nomex layer in material for additional chemical flash fire protection
Manufacturer: Trelleborg 477520066
 View more versions of this product
Catalog No. 19-810-028
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1991-2005
    Material Nomex™
    Includes Attached socks in the suit material, large, impact-resistant visor and anti-fog shield, ventilation system
    Color Red
    Size 3X-Large
  • Description & Specifications
Related Products
Product Content Feedback