Learn More
Ansell™ Trellchem™ Glove Systems Components
The TRELLCHEM™ glove assembly consists of several layers for maximum protection, chemical resistance and comfort. The gloves are attached with the TRELLCHEM™ Bayonet glove ring system, which offers quick and simple glove replacement.
Manufacturer:  Ansell™ 001230012
 View more versions of this product
Catalog No. 19-810-211
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Up to 4 layers of glove protection for use with Trellchem Gas-Tight and Splash Suits.
  • The inner chemically resistant silver glove is made of silver colored polymer barrier laminate
  • The inner glove is covered by a black outer glove made of special chloropene rubber
  • A Kevlar outer glove stretches to fit over the glove assembly to provide extra cut protection
  • A thin inner cotton layer can be added for extra comfort
Specifications
Specifications
Trelleborg™ Trellchem™ glove systems
Universal
Yellow
Dupont™ Kevlar™ aramid fibers
Cut-resistant Outer Glove