Learn More
Ansell™ Trellchem™ Glove Systems Components
The TRELLCHEM™ glove assembly consists of several layers for maximum protection, chemical resistance and comfort. The gloves are attached with the TRELLCHEM™ Bayonet glove ring system, which offers quick and simple glove replacement.
Specifications
For Use With (Equipment) Trelleborg™ Trellchem™ glove systems
View More Specs
Products 5
Catalog Number Mfr. No. Material Size Price Quantity & Availability  
Catalog Number Mfr. No. Material Size Price Quantity & Availability  
19-810-211 View Documents Ansell™
001230012
Dupont™ Kevlar™ aramid fibers Universal This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-207 View Documents Ansell™
072251210
PE and EVOH (SilverShield) 13 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-206 View Documents Ansell™
487010262
PE and EVOH (SilverShield) 11 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-810-205 View Documents Ansell™
487010261
PE and EVOH (SilverShield) 9 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-823-188 View Documents Ansell™
72250301
Viton™/Butyl Rubber Universal This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Up to 4 layers of glove protection for use with Trellchem Gas-Tight and Splash Suits.
  • The inner chemically resistant silver glove is made of silver colored polymer barrier laminate
  • The inner glove is covered by a black outer glove made of special chloropene rubber
  • A Kevlar outer glove stretches to fit over the glove assembly to provide extra cut protection
  • A thin inner cotton layer can be added for extra comfort
Specifications
Specifications
Trelleborg™ Trellchem™ glove systems