Learn More
Ansell™ Trellchem™ Level B evo Hazardous Protection Suits
Includes Nomex layer in material for additional chemical flash fire protection
Manufacturer:  Ansell™ 477320065
 View more versions of this product
Catalog No. 19-810-039
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Single skin; no overcover required
  • Antistatic resistance built into material; highly abrasion resistant
  • Large, impact-resistant visor and antifog shield
  • Available with attached socks in the suit material or attached nitrile black boots†
  • Includes ventilation system
  • Fully repairable
  • Red
Warranty and Services
Two years
Specifications
Specifications
EN 943-2, NFPA 1991
2X-Large
Larger visor
Nomex™
Red
Hazmat suit providing maximum protection against the most aggressive chemicals in all forms, including warfare agents
Attached socks