Trellchem™ Level B evo Hazardous Protection Suits Available on GSA/VA Contract for Federal Government customers only.

Includes Nomex layer in material for additional chemical flash fire protection

Manufacturer: Trelleborg 477320066BOOT

 View more versions of this product

Catalog No. 19-810-046

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance EN 943-2, NFPA 1991
    Material Nomex™
    Includes Attached Boots
    Color Red
    Size 3X-Large
    For Use With (Equipment) Hazmat suit providing maximum protection against the most aggressive chemicals in all forms, including warfare agents
    Style Larger visor

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit